Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD152 Hamster anti-Mouse, NA/LE, Unlabeled, Clone: 9H10, BD

Catalog No. BDB555278 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555278 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555278 Supplier BD Biosciences Supplier No. 555278

Hamster Monoclonal Antibody

The 9H10 antibody reacts with CD152 (CTLA-4), which is expressed on activated T lymphocytes 2-3 days after stimulation through T cell receptor. CTLA-4 has significant similarity to CD28 in amino acid sequence, structure, and genomic organization. Furthermore, CD152 and CD28 share common B7 family counter-receptors. Unlike CD28, CD152 expression at both mRNA and protein level appears to be restricted to activated T cells with a maximal surface expression between 48 and 72 hours post-activation. The regulation of CTLA-4 protein expression is complex and can involve changes in the rate of transit between intracellular stores and the cell surface. Whereas CD28 delivers a co-stimulatory signal required for T-cell activation, CTLA-4 is a negative regulator of cell-mediated immune responses. CD152 may play roles in induction and/or maintenance of immunological tolerance, regulation of protective immunity, and regulation of some aspects of thymocyte maturation. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leukocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD152
Applications Flow Cytometry
Classification Monoclonal
Clone 9H10
Concentration 1.0mg/mL
Conjugate Unconjugated
Description CTLA-4
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Hamster
Immunogen Mouse CTLA-4-human IgG1 fusion protein
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Bovine, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.