Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD154 (CD40L) Mouse anti-Human, APC, Clone: 89-76, BD

Catalog No. BDB648887 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB648887 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB648887 Supplier BD Biosciences Supplier No. 648887
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD154, clone 89-76, is derived from hybridization of mouse NS-1 cells with spleen cells from BALB/c mice immunized with sgp39-CD8 and activated human peripheral blood lymphocytes (PBLs).

CD154 (CD40 ligand or CD40L) recognizes a 39-kilodalton (kDa) antigen that is a member of the tumor necrosis factor (TNF) receptor family. CD154 is transiently expressed on activated CD4 T lymphocytes. The expression of CD154 by activated T-helper cells triggers B-cell cycling through binding to CD40. This co-receptor interaction is required for B-lymphocyte maturation, response to T-dependent ligands, and isotype switching. T-lymphocyte proliferation and generation of cytotoxic T lymphocytes (CTLs) is associated with the presence of CD154. Defects in the CD154 gene are associated with the X-linked hyper IgM syndrome.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD154
Applications Flow Cytometry
Classification Monoclonal
Clone 89-76
Concentration 100μg/mL
Conjugate APC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Activiate human peripheral blood lymphocytes (PBLs)
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.