Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD154 Mouse anti-Human, APC, Clone: TRAP1, BD

Catalog No. BDB555702 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555702 100 Tests
BDB560955 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555702 Supplier BD Biosciences Supplier No. 555702
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CD40 ligand (CD40L), a 39kDa type II membrane glycoprotein expressed on activated T cells. CD40-CD40L interaction plays a very important role in T cell-dependent B-cell proliferation, differentiation, and memory cell formation. Anti-CD40L mAb partially blocks T cell-B cell interaction affecting subsequent proliferation, differentiation, and memory cell formation. Anti-CD40L mAb partially blocks T cell-B cell interaction affecting subsequent proliferation, IL-2R expression and differentiation of B cells. In addition, blocking of CD40-CD40L interaction has been demonstrated with soluble CD40, resulting in the inhibition of immunoglobulin isotype switching. It has been reported that patients with X-linked hyper-IgM syndrome have defective expression of functional CD40L due to a defective gene that encodes CD40L. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: TRAP1
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD154
Applications Flow Cytometry
Classification Monoclonal
Clone TRAP1
Conjugate APC
Description CD40LG; CD40 ligand; CD40L; TNFSF5; T-BAM; TRAP
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.