Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD154 Hamster anti-Mouse, LE/AF (Low Endotoxin/Azide Free), Clone: MR1, Southern Biotech™

Hamster Monoclonal Antibody
Supplier: Southern Biotechnology Associates 165014
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD154 | |
Low Endotoxin/Azide-free (LE/AF) | |
0.5mg/mL | |
Hamster | |
0.5 mg | |
Primary | |
Store at -20°C |
Blocking Assay, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
MR1 | |
Supplied in PBS | |
Activated mouse Th1 clone D1.6 | |
RUO | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction