Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD154 Mouse anti-Human, NA/LE, Unlabeled, Clone: HCD40L-M90.1, BD

Catalog No. BDB552559 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552559 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552559 Supplier BD Biosciences Supplier No. 552559
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone hCD40L-M90.1 reacts with CD40 ligand (CD40L), a 39 kDa type II membrane glycoprotein expressed on activated T cells. CD40-CD40L interaction plays a very important role in T cell-dependent B-cell proliferation, differentiation, and memory cell formation. In addition, blocking of CD40-CD40L interaction has been demonstrated with soluble CD40, resulting in the inhibition of immunoglobulin isotype switching. It has been reported that patients with X-linked hyper-IgM syndrome have defective expression of functional CD40L due to a defective gene that encodes CD40L.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD154
Applications ELISA
Classification Monoclonal
Clone HCD40L-M90.1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CD40L
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.