Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD154 Mouse anti-Human, PE-Cy5, Clone: TRAP1 (also known as TRAP-1), BD

Catalog No. BDB561722 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561722 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561722 Supplier BD Biosciences Supplier No. 561722
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TRAP1 monoclonal antibody specifically binds to CD154. CD154 is a 39 kDa type II membrane glycoprotein that is a member of the tumor necrosis factor superfamily, Tumor necrosis factor ligand superfamily member 5 (TNFSF5). CD154 is expressed on a variety of cell types including activated CD4+ T cells and some CD8+ T cells, NK cells, mast cells and basophils. CD154 is also known as CD40 ligand (CD40L); it serves as a ligand for CD40 that is expressed on B cells, macrophages, and dendritic cells. The expression of CD154 by activated T-helper cells costimulates B-cell activation and proliferation through binding to CD40 expressed on B cells. In response to T-dependent antigens, the CD154 and CD40 interaction is required for B-lymphocyte differentiation, including immunoglobulin production and isotype switching and memory B cell generation. The TRAP1 antibody can partially block T cell-B cell interaction and inhibit the subsequent proliferation, differentiation, and memory formation of B cells. It has been reported that patients with X-linked hyper-IgM syndrome have defective expression of functional CD154 due to mutations in the CD40LG gene that encodes CD154.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD154
Applications Flow Cytometry
Classification Monoclonal
Clone TRAP1 (also known as TRAP-1)
Conjugate PE-Cyanine5
Description CD40LG; CD40 ligand; CD40L; TNFSF5; T-BAM; TRAP
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.