Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158a Mouse anti-Human, APC, Clone: HP-3E4, BD

Catalog No. BDB564319 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564319 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564319 Supplier BD Biosciences Supplier No. 564319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HP-3E4 monoclonal antibody recognizes CD158a. CD158a is a 58 kDa membrane glycoprotein, member of the killer-inhibitory receptor (KIR). The KIR family consists of transmembrane glycoproteins of the Ig superfamily expressed on <1-8% of NK cells and a subset of T cells which are involved in recognition of MHC class I molecules on target cells and inhibit cytotoxicity. CD158a is an antigen which recognizes HLA-C molecules (-Cw*0401 and -Cw*1503). This antibody is suitable for staining acetone-fixed, frozen tissue sections. It was reported that this clone also crossreacted with NKAT8 (CD158i) transfected cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158a
Applications Flow Cytometry
Classification Monoclonal
Clone HP-3E4
Conjugate APC
Description KIR2DL1; KI2L1; NKAT; NKAT-1; NKAT1; KIR-K64; KIR221; p58.1
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NK Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.