Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158a Mouse anti-Human, Unlabeled, Clone: HP-3E4, BD

Catalog No. BDB556061 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556061 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556061 Supplier BD Biosciences Supplier No. 556061
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts wth a 58 kDa membrane glycoprotein, member of the killer-inhibitory receptor (KIR). The KIR family family consists of transmembrane glycoproteins of the Ig superfamily expressed on <1-8% of NK cells and a subset of T cells which are involved in recognition of MHC class I molecules on target cells and inhibit cytotoxicity. CD158a is an antigen which recognizes HLA-C molecules (-Cw0401 and -Cw1503). This antibody the purified format is suitable for staining acetone-fixed, frozen tissue sections. It was reported that this clone also cross-reacted with NKAT8 (CD158i) tranfectant cells.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158a
Applications Flow Cytometry
Classification Monoclonal
Clone HP-3E4
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.