Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158b Mouse anti-Human, Unlabeled, Clone: CH-L, BD

Catalog No. BDB559783 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559783 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559783 Supplier BD Biosciences Supplier No. 559783
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CH-L monoclonal antibody specifically binds to CD158b proteins. These proteins are 50-58 kDa type I glycoproteins that belong to the killer cell inhibitory receptor (KIR) family. They are also known as CD158b1 (Killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 2/KIR2DL2; NKAT-6; p58.2); CD158b2 (Killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3/KIR2DL3; NKAT-2; p58.2); or CD158j (Killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2/KIR2DS2; NKAT-5; p50.2). CD158b molecules are composed of two extracellular Ig-like domains, a transmembrane region, and either long (76-84 amino acids) or short (39 amino acid) cytoplasmic tails. They are expressed on NK cells and subsets of TCR αβ+ cells or TCR γδ+ cells. Ligand- or CH-L antibody-bound CD158b1 or CD158b2 can reportedly inhibit cytolytic NK and T cell responses to various stimuli including certain target cells expressing MHC class I ligands encoded by HLA-C alleles (Cw 1, 3, 7 and 8). CD158j reportedly may enhance some cellular cytolytic responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158b
Applications Flow Cytometry
Classification Monoclonal
Clone CH-L
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.