Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15s Mouse anti-Human, Brilliant Violet 421, Clone: CSLEX1, BD

Catalog No. BDB563912 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563912 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563912 Supplier BD Biosciences Supplier No. 563912
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CSLEX1 monoclonal antibody specifically binds to CD15s. This anti-Sialyl-Le[x] monoclonal antibody is specific for the α 2-3 sialosylated form of lacto-N-fucopentaose III, sialyl Lex (Sle[x]). Sle[x] is expressed on granulocytes, monocytes and both normal and tumor cells of diverse origin. It has been shown to be a ligand for both endothelial leukocyte adhesion molecule-1 (ELAM-1 or E-selectin), and granule membrane protein-140 (GMP-140 or P-selectin).

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD15s
Applications Flow Cytometry
Classification Monoclonal
Clone CSLEX1
Conjugate Brilliant Violet 421
Description Sialyl Lewis X
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Membrane proteins from human tomach adenocarcinoma tissue
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.