Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/32 Rat anti-Mouse, LE/AF (Low Endotoxin/Azide Free), Clone: 93, Southern Biotech™
SDP

Catalog No. OB163014 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1630-14 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1630-14 Supplier Southern Biotechnology Associates Supplier No. 163014
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.

Specifications

Antigen CD16/32
Applications Blocking Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Low Endotoxin/Azide-free (LE/AF)
Clone 93
Concentration 0.5mg/mL
Formulation Supplied in PBS
Host Species Rat
Immunogen VH81X Tg B cells and T220 stromal cell line
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at -20°C
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.