Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD16/CD32 Rat anti-Mouse, FITC, Clone: 2.4G2, BD
Rat Monoclonal Antibody
$192.80 - $373.50
Specifications
Antigen | CD16/CD32 |
---|---|
Clone | 2.4G2 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561728
|
BD Biosciences
561728 |
0.1 mg |
Each for $192.80
|
|
|||||
BDB553144
|
BD Biosciences
553144 |
0.5 mg |
Each for $373.50
|
|
|||||
Description
The 2.4G2 antibody reacts specifically with a common nonpolymorphic epitope on the extracellular domains of the mouse FcγIII and FcγII receptors. It has also been reported to bind the FcγI receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the FcγIII and FcγII, and possibly FcγI, receptors in vitro and in vivo. CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.Specifications
CD16/CD32 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Mouse BALB/c Macrophage J774 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2.4G2 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
FcγRIII/FcγRII; Fcgr3/Fcgr2 |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title