Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/CD32 Rat anti-Mouse, PerCP-Cy5.5, Clone: 2.4G2, BD

Catalog No. BDB560540 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560540 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560540 Supplier BD Biosciences Supplier No. 560540
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 2.4G2 antibody reacts specifically with a common nonpolymorphic epitope on the extracellular domains of the mouse FcγIII and FcγII receptors. It has also been reported to bind the FcγI receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the FcγIII and FcγII, and possibly FcγI, receptors . CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16/CD32
Applications Flow Cytometry
Classification Monoclonal
Clone 2.4G2
Concentration 0.2mg/mL
Conjugate PerCP-Cyanine5.5
Description FcγRIII/FcγRII; Fcgr3/Fcgr2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse BALB/c Macrophage J774
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.