Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD16; Clone: 3G8; BD

Catalog No. BDB563785 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563785 100 Tests
BDB563784 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB563785 Supplier BD Biosciences Supplier No. 563785
Add to Cart
Add to Cart

Host Species: Mouse
Clone: 3G8
Isotype: IgG1, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Immunogen: Human polymorphonuclear leukocytes

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16
Classification Monoclonal
Clone 3G8
Conjugate Brilliant Ultraviolet 395
Host Species Mouse
Immunogen Human polymorphonuclear leukocytes
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.