Learn More
CD16 Mouse anti-Human, FITC, Clone: 3G8, BD
Mouse Monoclonal Antibody
$110.00 - $285.50
Specifications
Antigen | CD16 |
---|---|
Clone | 3G8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB560996
|
BD Biosciences
560996 |
25 Tests |
Each for $110.00
|
|
|||||
BDB556618
|
BD Biosciences
556618 |
50 Tests |
Each for $158.70
|
|
|||||
BDB555406
|
BD Biosciences
555406 |
100 Tests |
Each for $285.50
|
|
|||||
Description
Reacts with the 50-65kDa transmembrane form of IgG Fc (FcγRIII), a human NK-cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. CD16 antibody blocks the binding of soluble immune complexes to granulocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Mouse
Clone: 3G8
Isotype: IgG1, κ
Cross Reactivity: Baboon, Cynomolgus
Flow Cytometry
Specifications
CD16 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3 |
3G8 | |
Monoclonal | |
Mouse | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
Human polymorphonuclear leukocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.