Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16 Mouse anti-Human, PE, Clone: B73.1, BD

Catalog No. BDB561313 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561313 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB561313 Supplier BD Biosciences Supplier No. 561313
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD16 antibodies recognize an Mr 50- to 65kDa antigen present on human NK lymphocytes that is the IgG Fc receptor III.The CD16 antigen is expressed on approximately 15% of peripheral blood lymphocytes and is present on virtually all resting NK lymphocytes. The CD16 antigen can be expressed on CD3+ T lymphocytes from certain individuals. The CD16 antigen is also expressed on neutrophils. CD16 clone B73.1 reacts with neutrophils at a lower intensity than CD16 clone NKP15 and CD16 clone GO22. A variable number of CD16+ lymphocytes coexpress either the CD57 antigen or low-densityCD8 antigen or both.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16
Applications Flow Cytometry
Classification Monoclonal
Clone B73.1
Conjugate PE
Description IgG Fc receptor III; IGFR3; FCG3; FCGR3; FCGRIII; FcγRIII
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NK Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.