Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16 Mouse anti-Human, PE-Cy7, Clone: 3G8, BD

Catalog No. BDB560918 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
50 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560918 25 Tests
BDB560716 50 Tests
BDB557744 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BDB560918 Supplier BD Biosciences Supplier No. 560918
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the 50-65kDa transmembrane form of IgG Fc (FcγRIII), a human NK-cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. CD16 antibody blocks the binding of soluble immune complexes to granulocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 3G8
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16
Applications Flow Cytometry
Classification Monoclonal
Clone 3G8
Conjugate PE-Cyanine7
Description FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human polymorphonuclear leukocytes
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.