Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16 Mouse anti-Human, PE, Clone: 3G8, BD

Catalog No. BDB555407 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555407 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555407 Supplier BD Biosciences Supplier No. 555407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 3G8 monoclonal antibody specifically binds to the 50-65 kDa transmembrane form of the IgG Fc Receptor (FcγRIII), a human NK cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. The 3G8 antibody blocks the binding of soluble immune complexes to granulocytes. The 3G8 antibody is reported (Vossebeld et al., 1997) to increase intracellular calcium levels in human neutrophils by interacting with bothFcγRIIa and FcγRIIIb molecules. This antibody has also been reported to induce homotypic neutrophil aggregation.

This clone also cross-reacts with a subset of peripheral blood lymphocytes and monocytes, but not granulocytes, of baboon and both rhesus and cynomologus macaque monkeys. Multi-color analysis reveals that the distribution on lymphocytes is similar to that found in human studies with the majority of CD16-positive lymphocytes being both CD3 and CD20 negative.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16
Applications Flow Cytometry
Classification Monoclonal
Clone 3G8
Conjugate PE
Description FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human polymorphonuclear leukocytes
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.