Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD16 Mouse anti-Human, V450, Clone: 3G8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 644489
Description
The CD16 antibody, clone 3G8, is derived from the fusion of CD2F1 mouse spleen cells with P3U1 myeloma cells.
The CD16 antibodies recognize a 50 to 65-kilodalton (kDa) antigen present on human NK lymphocytes that is the IgG Fc receptor III.

Specifications
CD16 | |
Monoclonal | |
100μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human polymorphonuclear leukocytes | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
3G8 | |
Violet Fluor 450 (V450) | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction