Learn More
CD160 Mouse anti-Human, Alexa Fluor 488, Clone: BY55, BD
Catalog No. BDB562351 Shop All BD Biosciences Products
Change view
Click to view available options
100 Tests; Alexa Fluor 488
Catalog No.
BDB562351 100 Tests; Alexa Fluor 488
1 options
This item is not returnable. View return policy
Catalog No. BDB562351 Supplier BD Biosciences Supplier No. 562351
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mouse Monoclonal Antibody
The BY55 monoclonal antibody specifically binds to the human cell surface molecule, CD160 (also known as BY55). CD160 is a glycosyolphosphatidylinositol-anchored glycoprotein member of the immunoglobulin superfamily. It is expressed on cytotoxic αβ CD8+ T cells and natural killer (NK) cells, γδ T cells and intestinal intraepithelial T lymphocytes. CD160 can bind to classical and nonclassical MHC class I molecules. This interaction can costimulate the proliferation of activated human T lymphocytes including CD28-negative T lymphocytes.
TRUSTED_SUSTAINABILITY
WARNING: Reproductive Harm - www.P65Warnings.ca.gov