Learn More
CD160 Mouse anti-Human, Alexa Fluor 647, Clone: BY55, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562362
Description
The BY55 monoclonal antibody specifically binds to the human cell surface molecule, CD160 (also known as BY55). CD160 is a glycosyolphosphatidylinositol-anchored glycoprotein member of the immunoglobulin superfamily. It is expressed on cytotoxic αβ CD8+ T cells and natural killer (NK) cells, γδ T cells and intestinal intraepithelial T lymphocytes. CD160 can bind to classical and nonclassical MHC class I molecules. This interaction can costimulate the proliferation of activated human T lymphocytes including CD28-negative T lymphocytes.

Specifications
CD160 | |
Monoclonal | |
Alexa Fluor 647 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Human YT2C2 NK cell line | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
BY55 | |
BY55; BY55 antigen; NK1; NK28 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.