Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD160, Alexa Fluor 700, Clone: 688327, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6700N
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry
Specifications
CD160 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
BY55, BY55FLJ46513, CD160, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55, NK1, NK28 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11126 | |
Human | |
Purified |
Flow Cytometry | |
688327 | |
Flow Cytometry 5 uL/10^6 cells | |
O95971 | |
CD160 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD160 Ile27-Ser159 Accession # O95971 | |
100 Tests | |
Primary | |
Detects human CD160 in direct ELISAs. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD160 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction