Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD160 Mouse anti-Human, PE, Clone: BY55, BD

Catalog No. BDB562118 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562118 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562118 Supplier BD Biosciences Supplier No. 562118
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BY55 monoclonal antibody specifically binds to the human cell surface molecule, CD160 (also known as BY55). CD160 is a glycosyolphosphatidylinositol-anchored glycoprotein member of the immunoglobulin superfamily. It is expressed on cytotoxic αβ CD8+ T cells and natural killer (NK) cells, γδ T cells and intestinal intraepithelial T lymphocytes. CD160 can bind to classical and nonclassical MHC class I molecules. This interaction can costimulate the proliferation of activated human T lymphocytes including CD28-negative T lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD160
Applications Flow Cytometry
Classification Monoclonal
Clone BY55
Conjugate PE
Description BY55; BY55 antigen; NK1; NK28
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human YT2C2 NK Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.