Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD162 Mouse anti-Human, Unlabeled, Clone: KPL-1, BD

Catalog No. BDB556053 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556053 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556053 Supplier BD Biosciences Supplier No. 556053
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with p-selectin glycoprotein ligand-1 (PSGL-1), a 120 - 220kDa, mucin-like type I transmembrane glycoprotein. CD162 is expressed on neutrophils, monocytes, and most lymphocytes. CD162 binds to CD62P (P-Selectin), CD62E (E-Selectin) and CD62L (L-Selectin). This molecule mediates rolling of leukocytes on activated endothelium and the interaction between leukocytes and activated platelets or other leukocytes found at inflammatory sites. It has also been reported that this clone completely inhibits the interaction between P-Selectin and either CD4-T cells or neutrophils. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD162
Applications Flow Cytometry
Classification Monoclonal
Clone KPL-1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description PSGL-1, p-selectin glycoprotein ligand-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.