Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD162 Rat anti-Mouse, PE, Clone: 2PH1, BD

Catalog No. BDB555306 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555306 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555306 Supplier BD Biosciences Supplier No. 555306
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 2PH1 monoclonal antibody specifically binds to the N-terminus of CD162 (P-selectin glycoprotein ligand-1, PSGL-1), encoded by the Selplg gene. PSGL-1 is expressed on the cell surface as a homodimer of approximately 230 kDa. In the mouse, Selpl mRNA is detected in most tissues, with high levels found in hematopoietic cells, brain, and adipose tissue. Flow cytometric analyses have revealed CD162 expression on bone marrow-derived mast and dendritic cells, splenic leukocytes, platelets, peripheral blood neutrophils, and neutrophil and T-cell lines. PSGL-1 is a ligand for P-selectin (CD62P) and is involved in leukocyte rolling, the migration of leukocytes into inflamed tissues, and responses to vascular injury. It is a sialomucin that must be specifically sialylated, fucosylated, and sulfated to bind P-selectin. There is also evidence that other ligands for PSGL-1 and CD62P may exist. The 2PH1 antibody is reported to block binding of mouse leukocytes to CD62P, but the 4RA10 antibody (Cat. No. 557787) has significantly greater blocking activity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD162
Applications Flow Cytometry
Classification Monoclonal
Clone 2PH1
Concentration 0.2mg/mL
Conjugate PE
Description Selplg; PSGL-1; Psgl1; Selp1; Selpl; P-selectin glycoprotein ligand 1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Ovalbumin-conjugated peptide covering amino acids 42 to 60 of mouse PSGL-1
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.