Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD164 Mouse anti-Human, PE, Clone: N6B6, BD

Catalog No. BDB551298 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551298 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551298 Supplier BD Biosciences Supplier No. 551298

Mouse Monoclonal Antibody

Reacts with an 80-90 kDa, mucin-like molecule present on CD34+ cells during the early stages of B cell, erythroid cell and myelomonocytic cell development. It is also expressed on epithelial cells, peripheral blood monocytes and weakly on lymphocytes. Mucin-like molecules are a new family of glycoproteins present in tissues of the hematopoietic system. They are highly glycosylated polypeptides, containing predominantly O-linked carbohydrate side chains. Reports suggest that CD164 may play a role in hematopoiesis by facilitating the adhesion of CD34+ cells to bone marrow stroma.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD164
Applications Flow Cytometry
Classification Monoclonal
Clone N6B6
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Pre-B Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.