Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16b Mouse anti-Human, PE, Clone: CLB-GRAN11.5, BD

Catalog No. BDB550868 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550868 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550868 Supplier BD Biosciences Supplier No. 550868
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CD16b, a glycosyl phosphatidyl inositol-anchored (GPI) protein expressed on human neutrophils. Human CD16 is the low affinity Fc gamma receptor III (Fc gamma RIII) and shows two distinct forms coded by two linked genes. One form is CD16a, a polypeptide-anchored form (Fc gamma RIIIA), present on natural killer cells and macrophages. The other form is CD16b the GPI-anchored form, Fc gamma RIIIB, found on human neutrophils. CD16b is polymorphic and the two codominant alleles are referred to as Neutrophil Antigen 1 (NA1) and Neutrophil Antigen 2 (NA2). Clone CLBgran11.5 reacts with neutrophils expressing the NA1 molecule. CD16b has been reported to participate in immune complex binding by resting neutrophils and also inneutrophil transendothelial migration by interacting with integrins during the inflammation response process.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16b
Applications Flow Cytometry
Classification Monoclonal
Clone CLB-GRAN11.5
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.