Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD171 Mouse anti-Human, PE, Clone: 5G3, BD

Catalog No. BDB564193 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564193 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564193 Supplier BD Biosciences Supplier No. 564193

Mouse Monoclonal Antibody

Neurite adhesion molecule L1 has been implicated in neuron-neuron and neuron-Schwann cell adhesion in vertebrates. L1-like molecules, found in mouse, rat, chicken, and human, promote axonal elongation and may also play a role in regeneration of axons after injury. Molecular cloning data suggest 87% amino acid identity between mouse and human L1 molecules. 5G3 antigen (Ag), originally defined by monoclonal antibody 5G3, is considered to be the human homologue of mouse L1. The 5G3 antibody was developed against a human neuroblastoma cell line to use as a probe for the elucidating the biological characteristics of neuroblastoma. 5G3 specifically recognizes a neuroblastoma target glycoprotein antigen of 215 kDa and its 200 kDa precursor. The 215 kDa molecule is expressed on the cell surface; whereas the 200 kDa precursor is shed from the cell surface. The 215 and 200 kDa species also differ in their posttranslational modification patterns. The 5G3 antibody has been used as a marker for neuroblastoma, and to purify 5G3 Ag from normal adult human brain.

The antibody recognizes human L1 on human neuroblastoma cell lines and tissues. Reactivity has been tested on a variety of malignant and normal tissues. Squamous lung, squamous skin, and osteogenic sarcoma cell lines were positive, as were two out of eight melanoma cell lines tested. A variety of other cell lines and tumor tissues tested negative. 5G3 did not react with either T or B lymphoblastoid cell lines or a fibroblast cell line. Among all the normal tissues tested, mAb 5G3 reacted only with cerebellum.

The molecular masses observed using mAb 5G3 may vary among immunoprecipitation isolates. In normal human cerebellum, 5G3 Ag migrated as a 190/200 kDa doublet, 140 kDa band with minor bands at 80 and 65 kDa. 5G3 Ag isolated from SK-N-AS cells migrates as 200 to 215 kDa bands, or as a diffuse band ranging from 200 to 215 kDa. Additional bands have been described at 140 to 150 kDa in SK-N-AS cells. Only the 200 kDa band has been detected in culture media from SK-N-AS cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD171
Applications Flow Cytometry
Classification Monoclonal
Clone 5G3
Conjugate PE
Description L1 Neuite Cell Adhesion Molecule
Host Species Mouse
Immunogen Human SK-N-AS Neuroblastoma Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.