Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD172a/b Mouse anti-Human, Alexa Fluor 647, Clone: SE5A5, BD

Catalog No. BDB565035 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565035 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565035 Supplier BD Biosciences Supplier No. 565035
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SE5A5 monoclonal antibody specifically binds to a common epitope on CD172a/ SIRPα (90 kDa) and CD172b/ SIRPβ 1(50 kDa) . These transmembrane glycoproteins are members of the Signal Regulatory Protein (SIRP) family that, in turn, belongs to the Immunoglobulin superfamily. The SIRP family is comprised of two subgroups, SIRPα and SIRPβ that are distinguished by the presence (α) or absence (β) of a cytoplasmic domain containing two immunoreceptor tyrosine-based inhibition motifs (ITIM). CD172a/ SIRPα is expressed on CD34+ stem/progenitor cells, cardiomyocytes, monocytes, macrophages, granulocytes, dendritic cells, and in the central nervous system. It binds to CD47 and is implicated in mediating inhibitory signals via the ITIM/SHP-2 association. CD172b/ SIRPβ to cellular activation signaling . CD172b is expressed on myeloid cells, including peripheral blood monocytes and granulocytes. It is not expressed on CD34+ cells. CD172a and CD172b have complementary role s in signal regulation and may work together in tuning certain cellular responses to stimuli.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD172a/b
Applications Flow Cytometry
Classification Monoclonal
Clone SE5A5
Conjugate Alexa Fluor 647
Description SIRP alpha/beta1; SIRPα/SIRPβ1; Signal Regulatory Protein α/β1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Host Species Mouse
Immunogen Human SIRP alpha extracellular domain Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.