Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD172b Mouse anti-Human, PE, Clone: B4B6, BD

Catalog No. BDB552602 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552602 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552602 Supplier BD Biosciences Supplier No. 552602
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 50 kDa, type I transmembrane glycoprotein known as signal regulatory protein β (SIRPβ). The SIRPs are members of the immunoglobulin superfamily. SIRPβ does not possess a cytoplasmic domain but instead the transmembrane domain contains a positively-charged residue that interacts with one other transmembrane protein (DAP-12 or KARAP). DAP-12 has two immunoreceptor tyrosine-based activation motifs (ITAM) within its cytoplasmic domain that are thought to link SIRPβ to activating machinery. SIRPβ is expressed on peripheral blood monocytes and granulocytes, but is not expressed on CD34+ cells. SIRPα and SIRPβ have complementary roles in signal regulation and may work together in tuning the response to a stimulus

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD172b
Applications Flow Cytometry
Classification Monoclonal
Clone B4B6
Conjugate PE
Description SIRPB1; SIRP-BETA-1; SIRPβ1; SIRPbeta; SIRP beta
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human SIRPβ1 extracellular domain Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.