Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD177 Mouse anti-Human, Brilliant Violet 421, Clone: MEM-166, BD

Catalog No. BDB564240 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564240 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564240 Supplier BD Biosciences Supplier No. 564240
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MEM-166 monoclonal antibody specifically binds to CD177 which is also known as NB1 (Neutrophil-specific antigen), HNA-2a (Human neutrophil alloantigen 2a), and PRV1 (Polycythemia rubra vera 1). CD177 is a glycophosphatidylinositol (GPI)-anchored plasma membrane glycoprotein (56-64 kDa). CD177 is expressed on subpopulations of neutrophils, neutrophillic metamyelocytes and myelocytes from 89 to 97% of healthy individuals. It is also expressed on secondary granules. Its expression is upregulated on granulocytes stimulated with the chemotactic peptide f-met-leu-phe (fMLP). Although the function of CD177 has not been fully established, some reports suggest its possible role as receptor molecule involved in leucocyte migration.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD177
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-166
Conjugate Brilliant Violet 421
Description NB1
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.