Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD178 Hamster anti-Rat/Mouse, NA/LE, Unlabeled, Clone: MFL4, BD

Catalog No. BDB555021 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555021 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555021 Supplier BD Biosciences Supplier No. 555021

Hamster Monoclonal Antibody

The MFL4 antibody, generated against mouse CD178 (Fas Ligand, CD95 Ligand), cross-reacts with the rat Fas Ligand antigen. MFL4 mAb detects rat Fas Ligand expressed by transfected cells, but not the parental cell line. In addition, the epitope detected by MFL4 can be induced on activated rat splenic T lymphocytes. The MFL4 mAb has been reported to inhibit killing of mouse lymphoma line A20.2J by rat FasL-transfected cells. The tissue distribution of mouse Fas Ligand has been well characterized. It is expressed at the mRNA and protein levels in a variety of tissues and cell types, including spleen, bone marrow, testis, uterus, eye, and activated lymphocytes. The expression of Fas Ligand in the rat has been detected in liver Kupffer cells, T cells in the central nervous system, spleen, and corpus luteum. MFL4 antibody reacts with the mouse Fas Ligand protein in a pattern similar to that of MFL3 antibody. Fas Ligand is a type-II transmembrane protein, a member of the TNF/NGF superfamily and a ligand for Fas (CD95). Fas Ligand and its counter- receptor CD95 are believed to participate in T-cell development, the regulation of immune responses, and cell-mediated cytotoxic mechanisms. It has been reported that human Fas Ligand is released from the surface of activated lymphocytes by metalloproteinase.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD178
Applications Flow Cytometry
Classification Monoclonal
Clone MFL4
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Fas Ligand, CD95 Ligand
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm filtered. Endotoxin level is ≤0.01 ng/μg of protein.
Host Species Hamster
Immunogen Mouse FasL transfected cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG κ3
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.