Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD179b Rat anti-Mouse, Biotin, Clone: LM34, BD

Catalog No. BDB551865 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551865 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551865 Supplier BD Biosciences Supplier No. 551865
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The pre-B cell receptor (pre-BCR) expressed during the early stages of B lymphocyte development is a heterodimer of immunoglobulin heavy chain (IgH) with surrogate light chain, which is an Ig-light-chain-like molecule composed of the non-covalently linked CD179b (μ5) and CD179a (VpreB) proteins. The pre-BCR is believed to control IgH repertoire selection and proliferation of differentiating B lymphocytes. The LM34 antibody reacts with μ5 and surrogate light chain, but not VpreB alone, in transfected X63-Ag8.653 cells. It detects surrogate light chain on pro-B and pre-B cell lines, in the presence or absence of IgH, but not on IgM-positive B lymphocytes. It also detects surrogate light chain or μ5 on the surface and in the cytoplasm of pro-B or pre-B cells from the bone marrow of normal or RAG2-deficient mice. It has been noted that the (CD45R/B220) cell-surface expression of surrogate light chain is upregulated after a one-hour incubation of bone-marrow leukocytes in tissue culture medium at 37°C. At the earliest stages of B lymphopoiesis, before IgH is available, the surrogate light chain associates with a complex of glycoproteins, including a nonclassical cadherin, which could be involved in selective adhesion events during B-lymphocyte development. After immunization, cell-surface μ5 is detected on a subset of splenic Ig light chain-positive germinal-center B lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD179b
Applications Flow Cytometry
Classification Monoclonal
Clone LM34
Concentration 0.5mg/mL
Conjugate Biotin
Description λ5
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Purified soluble complex of recombinant μ IgH, λ5, and V[preB]
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.