Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18 Rat anti-Mouse, Unlabeled, Clone: GAME-46, BD

Catalog No. BDB557440 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557440 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557440 Supplier BD Biosciences Supplier No. 557440
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The GAME-46 antibody reacts with the common β2 chain of LFA-1 (CD11a/CD18, αLβ2 integrin), Mac-1 (CD11b/CD18, αMβ2 integrin), and gp150, 95 (CD11c/CD18, αXβ2 integrin). Expression of CD18 is limited to leukocytes, where it is widely distributed in consort with each of the three integrin α chains (CD11a, CD11b, and CD11c). Among splenocytes, NK cells have the highest density of CD18, and T lymphocytes express a higher density than the remaining cells. The β2 integrins are important mediators of leukocyte-endothelium interactions. It has been reported that GAME-46 antibody blocks in vitro adhesion of LFA-1-expressing cells to the ligands ICAM-1, ICAM-2 and ICAM-3 and of Mac-1-expressing cells to ICAM-1, C3bi, and fibrinogen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Applications Flow Cytometry
Classification Monoclonal
Clone GAME-46
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Integrin β2 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen LFA-1 (CD11a/CD18) from mouse T-cell hybridoma TAM8C41
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.