Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD18 Clone: IB4,

Catalog No. 217660100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
21-766-0100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-766-0100UG Supplier MilliporeSigma™ Supplier No. 217660100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD18 Clone: IB4 in Human samples, and it is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Inhibition Assays

Human CD18 (β2 integrin) is expressed on leukocytes as part of a heterodimeric complex with either CD11a, b or c (integrins αL, αM, or αX). CD18/CD11 heterodimers function in cell adhesion in immune and inflammatory responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD18
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone IB4
Conjugate Unconjugated
Formulation In 150mM NaCl, 100mM KCl, 50mM sodium phosphate, pH 7.5.
Host Species Mouse
Immunogen human peripheral blood monocytes
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽95kDa β2 integrin subunit. Blocks the binding of ICAM-1 and ICAM-3 to LFA-1.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.