Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD18 Rat anti-Mouse, Unconjugated, Clone: C71/16, Southern Biotech™

Rat Monoclonal Antibody
Supplier: Southern Biotechnology Associates 157001
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
| CD18 | |
| Monoclonal | |
| 0.5mg/mL | |
| Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added | |
| Cell membrane glycoproteins derived from mouse BW5147 T cell lymphoma cells | |
| RUO | |
| Mouse | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| C71/16 | |
| Unconjugated | |
| Rat | |
| 0.5 mg | |
| Primary | |
| Store at 2-8°C | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction