Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD18 (Integrin Beta-2) Ab-1, Clone: 68-5A5, Epredia™ LabVision

Catalog No. MS655P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS655P 1 mL 200μg/mL
MS655P0 100 μL 200μg/mL
MS655P1 500 μL 200μg/mL
MS655P1ABX 100 μL 1mg/mL
MS655PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS655P Supplier Epredia Supplier No. MS655P

Mouse Monoclonal Antibody

CD18 or integrin Β-2 is a transmembrane glycoprotein. It complexes non-covalently with either α L, α M, or α X integrin (CD11a, b, or c) to form the heterodimers, LFA-1, Mac-1, and p150,95, respectively. LFA-1 is the receptor for three members of the Ig supergene family of proteins, ICAM-1 (CD54), ICAM-2 (CD102), and Mac-1 and p150,95 bind to ICAM-1, fibrinogen, and iC3b. ICAM-3 (CD50). CD18/CD11 heterodimeric molecules are involved with cell/cell and cell/extracellular adhesion in immune and inflammatory responses.

Host Species: Mouse
Clone: 68-5A5
Isotype: IgG2a
Cross Reactivity: Human. Others-not tested.
Epitope: Not determined
Immunogen: Stimulated human leukocytes.
Molecular Weight: 95kDa
Positive Control: Tonsil.
Cellular Localization: Cell membrane

Flow Cytometry, Immunofluorescence, Immunohistochemistry

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen CD18 Ab-1
Classification Monoclonal
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Stimulated human leukocytes.
Purification Method Tonsil
Quantity 1 mL
Primary or Secondary Primary
Target Species Human
Content And Storage 2–8 degrees
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.