Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD180 Rat anti-Mouse, Biotin, Clone: RP/14, BD

Catalog No. BDB552129 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552129 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552129 Supplier BD Biosciences Supplier No. 552129

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

The RP/14 antibody reacts with CD180, the “radioprotective” 105 kDa (RP105) antigen expressed on mature B lymphocytes. Like other members of the Toll-like Receptor (TLR) family, the extracellular region of CD180 contains tandem repeats of a leucine-rich motif flanked by regions containing conserved cysteines. These characteristic structural components of TLR may mediate protein-protein interactions and regulation of signal transduction involved in innate immune responses. Furthermore, CD180 associates with the secretory protein MD-1 on the cell surface. Cross-linking of cell-surface CD180 with the RP/14 antibody induces activation signals in B lymphocytes. Despite the expression of CD180 on lymphocytes of all strains tested (BALB/c, BALB/c.xid, C3H, C57BL/6, CBA/N, and 129/Sv), some strains (BALB/c.xid, C57BL/6.xid, CBA/N, 129/Sv, and 129/Ola) display deficient responses to that stimulatory effects of mAb RP/14.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD180
Applications Flow Cytometry
Classification Monoclonal
Clone RP/14
Concentration 0.5mg/mL
Conjugate Biotin
Description Ly78; Ly-78; Lymphocyte antigen 78; RP105; Radioprotective 105 kDa protein
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c mouse splenocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.