Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD180 Mouse anti-Human, PE, Clone: G28-8, BD

Catalog No. BDB551953 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551953 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551953 Supplier BD Biosciences Supplier No. 551953
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with RP105/Bgp95, a 95-105 kDa type I membrane protein consisting of extracellular leucine-rich repeats and a short cytoplasmic domain. It is expressed on mantle zone B cells, but weakly on germinal center B cells. RP105/Bgp95 is also expressed on peripheral blood monocytes, dendritic cells, and a subset of peripheral blood lymphocytes. The extracellular domain associates with a molecule called MD-1 to form a cell surface receptor complex RP105/Bgp95/MD- 1. This receptor belongs to the family of toll-like receptors (TLR). Studies show that RP105/Bgp95/MD-1, working in concert with TLR4, controls B cell recognition and signaling of lipopolysaccharide (LPS). Reports on functional studies show that G28-8 monoclonal antibody can induce a G0 to G1 cell cycle transition and was synergistic with PMA, anti-µ, or anti-CD40 in inducing proliferation of resting B cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD180
Applications Flow Cytometry
Classification Monoclonal
Clone G28-8
Conjugate PE
Description RP105
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.