Learn More
CD184 Rat anti-Mouse, APC, Clone: 2B11/CXCR4, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 558644
Description
The 2B11/CXCR4 monoclonal antibody specifically reacts with mouse CD184, which is also known as CXC chemokine receptor, CXCR4. CXCR4 (previously known as Fusin and LESTR), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CXC chemokines, SDF-1/CXCL12. Mouse CXCR4 shows 91% homology at amino acid level with human CXCR4. CXCR4 is widely expressed by hematopoietic and non-hematopoietic cell types including neutrophils, monocytes, T cells, B cells, CD34-positive progenitor cells, endothelial cells, neurons and astrocytes. In the thymus CXCR4 is restricted to CD4+CD8+ cells, while in the spleen, predominant expression is found on B lymphocytes. Human CXCR4 is used by T-tropic HIV-1 as a co-receptor for viral entry. The mouse CXCR4 gene has been mapped to chromosome 1.

Specifications
CD184 | |
Monoclonal | |
0.2mg/mL | |
CXCR4, C-X-C chemokine receptor type 4; Fusin; LESTR; PB-CKR; Sdf1r | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
2B11/CXCR4 | |
APC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
GST-NCXCR4 fusion protein | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.