Learn More
CD184 Rat anti-Human, Unlabeled, Clone: 1D9, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 551413
Description
The monoclonal antibody 1D9 reacts with the human CD184 which is also known as CXC chemokine receptor, CXCR4. CXCR4 (previously known as fusin, LESTR and HUMSTR), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CXC chemokines, SDF-1/CXCL12. CXCR4 was widely expressed by hematopoietic and non-hematopoietic cell types including PMN, monocytes, T cells, B cells, CD34+ progenitor cells, endothelial cells, neurons and astrocytes. The human CXCR4 is used by T-tropic HIV-1 as a co-receptor for viral entry. The human CXCR4 gene has been mapped to chromosome 2q21. The immunogen used to generate 1D9 hybridoma was human CXCR4 fusion protein.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD184 | |
Monoclonal | |
0.5mg/mL | |
CXCR4, Fusin | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
1D9 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CXCR4 fusion protein | |
0.1 mg | |
Immunology | |
Human | |
IgG2a κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.