Learn More
CD184 Rat anti-Mouse, Unlabeled, Clone: 2B11/CXCR4, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 551852
Description
The 2B11/CXCR4 monoclonal antibody specifically binds to mouse CD184 which is also known as the CXC chemokine receptor, CXCR4. CXCR4 (previously known as Fusin and LESTR), is a seven-transmembrane, G-protein-coupled receptor. It is the specific receptor for the CXC chemokine, SDF-1/CXCL12. Mouse CXCR4 shows 91% homology at the amino acid level with human CXCR4. CXCR4 is widely expressed by hematopoietic and non-hematopoietic cell types including neutrophils, monocytes, T cells, B cells, CD34-positive progenitor cells, endothelial cells, neurons and astrocytes. Human CXCR4 is used by T-tropic HIV-1 as a co-receptor for viral entry. The mouse Cxcr4 gene has been mapped to chromosome 1.

Specifications
CD184 | |
Monoclonal | |
0.5mg/mL | |
CXCR4, C-X-C chemokine receptor type 4; Fusin; LESTR; PB-CKR; Sdf1r | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
2B11/CXCR4 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
GST-NCXCR4 fusion protein | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.