Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD184 Mouse anti-Human, PE-CF594, Clone: 12G5, BD

Catalog No. BDB562389 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562389 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562389 Supplier BD Biosciences Supplier No. 562389
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 12G5 monoclonal antibody specifically binds to CD184, also known as CXCR4 and Fusin. CD184/CXCR4 is a seven-transmembrane domain, G-protein-linked, glycoprotein chemokine receptor. CD184 serves as a receptor for the C-X-C chemokine, SDF-1. It is expressed on a wide variety of hematopietic cells, vascular endothelial cells and cells of the nervous system. CD184 plays a variety of roles in hematopoiesis, vascularization and neural development. CD184 also functions as a coreceptor for infection with T-cell tropic strains of HIV-1 and as a receptor for CD4-independent infection by some HIV isolates. The 12G5 antibody has been reported to block CD4-independent infection by HIV-2 and CD4-dependent infection by some T-cell tropic isolates of HIV-1.

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD184
Applications Flow Cytometry
Classification Monoclonal
Clone 12G5
Conjugate PE-CF594
Description CXCR4; Fusin; SDF-1 receptor; LAP3; LCR1; LESTR; NPYY3R; NPY3R; WHIM; HM89
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen SIVmac variant CP-MAC-infected Sup-T1 cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.