Learn More
CD19, Mouse, Clone: 1C9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CD19.
Supplier: Abnova Corporation H00000930M02
Description
Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. This gene encodes a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. [provided by RefSeq
Sequence: QPGPPSEKAWQPGWTVNVEGSGELFRWNVSDLGGLGCGLKNRSSEGPSSPSGKLMSPKLYVWAKDRPEIWEGEPPCVPPRDSLNQSLSQDSpecifications
CD19 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001770 | |
CD19 | |
CD19 (NP_001761, 98 a.a. ∼ 187 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunofluorescence, Western Blot | |
1C9 | |
Mouse monoclonal antibody raised against a partial recombinant CD19. | |
CD19 | |
B4/MGC12802 | |
Mouse | |
Affinity chromatography | |
RUO | |
930 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.