Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Rat anti-Mouse, Unconjugated, Clone: 6D5, Southern Biotech™

Rat Monoclonal Antibody
Supplier: Southern Biotechnology Associates 157501
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD19 | |
Monoclonal | |
0.5mg/mL | |
Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added | |
Mouse CD19-expressing K562 human erythroleukemia cells | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
6D5 | |
Unconjugated | |
Rat | |
0.5 mg | |
Primary | |
Store at 2-8°C | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction