Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Magnetic Particles - DM Mouse anti-Human, BD IMag-DM, Clone: 4G7, BD

Catalog No. BDB558497 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558497 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558497 Supplier BD Biosciences Supplier No. 558497
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BD IMag™ anti-human CD19 Particles - DM are magnetic nanoparticles that have monoclonal antibody conjugated to their surfaces. These particles are optimized for the positive selection or depletion of CD19-bearing leukocytes using the BD IMag™ Cell Separation Magnet (Cat. No. 552311).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD19
Applications Flow Cytometry
Classification Monoclonal
Clone 4G7
Conjugate BD IMag-DM
Host Species Mouse
Immunogen Human chronic lymphocytic leukemia (CLL) cells
Quantity 5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage The antibody-conjugated magnetic particles should be stored undiluted at 4°C.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.