Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Rat anti-Mouse, FITC, Clone: 6D5, MilliporeSigma™

Catalog No. FCMAB218FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB218FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB218FMI Supplier MilliporeSigma Supplier No. FCMAB218F

Rat Monoclonal Antibody

Detect CD19 using this Milli-Mark™ Anti-CD19-FITC Antibody, clone 6D5 validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD19
Applications Flow Cytometry
Classification Monoclonal
Clone 6D5
Conjugate FITC
Formulation Purified rat monoclonal IgG2ak conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_033974
Gene Symbols Cd19
Host Species Rat
Immunogen Full-length murine CD19 cDNA cloned into a vector and expressed in K562 human erythroleukemia cells.
Purification Method Protein G Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.