Learn More
CD19 Mouse anti-Human, PerCP-Cy5.5, Clone: SJ25C1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340951
Description
The CD19 antibody, clone SJ25C1, is derived from the hybridization of Sp2/0 mouse myeloma cells with spleen cells isolated from BALB/c mice immunized with NALM1 and NALM16 cells.
The CD19 antibody recognizes a 90-kilodalton (kDa) antigen that is present on human B lymphocytes.

Specifications
CD19 | |
Monoclonal | |
5μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human NALM1 + NALM16 cells | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
SJ25C1 | |
PerCP-Cyanine5.5 | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.