Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Mouse anti-Human, V450, Clone: HIB19, BD

Catalog No. BDB560354 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
120 Tests
30 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560354 30 Tests
BDB560353 120 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560354 Supplier BD Biosciences Supplier No. 560354
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the 95kDa type I transmembrane glycoprotein expressed during all stages of B-cell differentiation and maturation, except on plasma cells. CD19 is also present on follicular dendritic cells. It is not found on T cells or on normal granulocytes. CD19 plays a role in regulation of B-cell proliferation. It associates with the complement receptor 2 (CD21), TAPA-1 (CD81), Leu 13, and/or MHC class II to form a signal transduction complex on the surface of B cells. Anti-CD19 clone HIB19 partially blocks clone B43, another C19 mAb. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Mouse
Clone: HIB19
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD19
Applications Flow Cytometry
Classification Monoclonal
Clone HIB19
Conjugate Violet Fluor 450 (V450)
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 30 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.