Learn More
CD193 Rat anti-Mouse, Alexa Fluor 647, Clone: 83103, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 557974
Description
The 83103 monoclonal antibody specifically binds to CD193, which is also known as, C-C chemokine receptor type 3 (CCR3). CD193 is a chemokine receptor for CCL11/eotaxin, CCL24/eotaxin-2, CCL26/eotaxin-3, CCL5/RANTES, CCL7/MCP-3, CCL8/MCP-2, CCL14/MCP-4, CCL15/HCC-2 and CCL3/MIP-1a. CD193 is reportedly expressed on eosinophils, mast cells and Th2 cells. It plays an important role in eosinophil recruitment to the skin and the lung and in the development of airway hyperresponsiveness.

Specifications
CD193 | |
Monoclonal | |
0.2mg/mL | |
CCR3; CCR-3; chemokine (C-C) receptor 3; CC-CKR3; C-C CKR-3; CKR3; CC-CKR-3 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
83103 | |
Alexa Fluor 647 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse CCR3 transfected Cells Transfected Cell Line | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2a |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.